Please login to your online account to display your discounted pricing
Corning™ BioCoat™ Collagen I Multiwell Plates
Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
$156.51 - $5,881.77
Specifications
Lid Yes
Sterile No
Surface Coating ECM/Attachment Factors
Certifications/Compliance ANSI, SBS
Material Polystyrene
View More Specs
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products
Catalog Number Mfr. No. Quantity Color Type Well Color Price Quantity    
08-775-32
corning™
C354397
5/Cs. Black 384-Well Collagen I Multiwell Microplate, Low Volume Clear Case of 5 Each for $405.88
08-774-314
corning™
C354665
5/Cs. White 384-Well Collagen I Multiwell Microplate Opaque Case of 5 Each for $308.74
08-774-317
corning™
C356650
50/Cs. White 96-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $2,913.67
08-774-324
corning™
C356665
50/Cs. White 384-Well Collagen I Multiwell Microplate Opaque Case of 50 Each for $2,434.92
08-772-71
corning™
C356408
50/Cs. Clear 24-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,383.72
08-772-70
corning™
C356500
50/Cs. Clear 12-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,283.64
08-772-72
corning™
C356407
50/Cs. Clear 96-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,469.54
08-774-1
corning™
C354400
5/Cs. Clear 6-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $156.51
08-774-2
corning™
C354500
5/Cs. Clear 12-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $221.26
08-774-266
corning™
C356505
50/Cs. Clear 48-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,813.74
08-774-294
corning™
C354519
5/Cs. White 96-Well Collagen I Multiwell Microplate Opaque Case of 5 Each for $248.67
08-774-297
corning™
C356519
50/Cs. White, Opaque 96-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,700.46
08-774-299
corning™
C356649
50/Cs. Black 96-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $2,740.95
08-774-3
corning™
C354408
5/Cs. Clear 24-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $162.61
08-774-307
corning™
C354650
5/Cs. White 96-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $399.90
08-774-313
corning™
C354664
5/Cs. White 384-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $502.62
08-774-316
corning™
C354667
5/Cs. Black 384-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $490.28
08-774-325
corning™
C356666
50/Cs. Clear 384-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $2,324.54
08-774-4
corning™
C354505
5/Cs. Clear 48-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $306.95
08-774-5
corning™
C354407
5/Cs. Clear 96-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $167.54
10-1300
corning™
C356699
80/Cs. White 96-Well Collagen I Multiwell Microplate Opaque Case of 80 Each for $2,514.10
10-1302
corning™
C356701
80/Cs. White 96-Well Collagen I Multiwell Microplate Opaque Case of 80 Each for $4,311.10
10-1301
corning™
C356700
80/Cs. Black 96-Well Collagen I Multiwell Microplate Clear Case of 80 Each for $4,311.10
10-1306
corning™
C356705
80/Cs. Black 384-Well Collagen I Multiwell Microplate Clear Case of 80 Each for $5,881.77
08-772-69
corning™
C356400
50/Cs. Clear 6-Well Collagen I Multiwell Microplate Clear Case of 50 Each for $1,556.18
08-774-296
corning™
C354649
5/Cs. Black 96-Well Collagen I Multiwell Microplate Clear Case of 5 Each for $355.51
10-1299
corning™
C356698
80/Cs. Clear 96-Well Collagen I Multiwell Microplate Clear Case of 80 Each for $2,302.74
15-100-160
corning™
C4582
10/Cs. Half Area Black 96-Well Half Area Microplate Clear Case of 10 Each for $1,787.97
Description & Specifications
Specifications
Lid Yes
Sterile No
Surface Coating ECM/Attachment Factors
Certifications/Compliance ANSI, SBS
Material Polystyrene
  • Source: Rat tail
  • TC-treated
  • Clear, round well
  • Flat bottom
  • Nonsterile
For research use only.
Safety and Handling
6 Months