Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Collagen I Multiwell Plates


Treated with BioCoat™ for enhanced cell attachment and growth of a variety of primary cells and ransformed cell lines in serum-free or reduced serum cultures. Corning™ BioCoat™ Collagen I Multiwell Plates closely mimmics in vivo environments for cell culture applications. Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.

Manufacturer: corning™  C356650

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-317

  • / Case of 50 Each
 

Description & Specifications

Specifications

Lid Yes
Culture Area 0.32cm2
For Use With (Application) Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Format 96-well
Material Polystyrene
Quantity 50/Cs.
Shelf Life 6 Months
Source Rat Tail
Sterile No
Storage Requirements 4° to 30°C
Surface Coating ECM/Attachment Factors
Type 96-Well Collagen I Multiwell Microplate
Certifications/Compliance ANSI, SBS
Well Color Clear
Well Design Round
Closure Yes
Color White
Well Shape Round
Bottom Plate Material Polystyrene
Well Color Clear
Closure Yes
Applications Promotion of cell attachment and spreading, rapid expansion of cell populations, serum-free or reduced serum culture, cell adhesion assay, improving survival of primary cells in culture
Surface Coating ECM/Attachment Factors

  • Source: Rat tail
  • TC-treated
  • Clear, round well
  • Flat bottom
  • Nonsterile

For research use only.