Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Collagen I Inserts with PET Membrane


Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Manufacturer: corning™  C354442

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-774-18

  • / Case of 24 Each
 

Description & Specifications

Specifications

Inserts per Plate 6
Quantity 4
No. of Wells 6
Pore Size 0.45μm
For Use With (Application) Adhesion, growth and differentiation of neuronal and transfected cell lines
Storage Requirements 2° to 8°C
Material PET
Type Well Plate