Please login to your online account to display your discounted pricing

Corning™ BioCoat™ Assay System


Automation-friendly design

Manufacturer: corning™  C354804

 View more versions of this product

 Alert

In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.

Catalog No. 08-788

  • / Pack of 5 Each
 

Description & Specifications

Specifications

Lid Yes
For Use With (Equipment) Falcon™ companion plate
Format 24-well
Quantity 5/Cs.
Includes 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid
Surface Coating Fibrillar Collagen
No. of Wells 24
Pore Size 1μm
Closure Yes
Membrane Polyethylene Terephthalate
Culture Area 0.3cm2
Color Transparent
Sterile No
Type 24 Multiwell Insert Plate