Please login to your online account to display your discounted pricing
Corning™ BioCoat™ Assay System
Automation-friendly design
Manufacturer: corning™  C354801
 View more versions of this product
 Alert
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Catalog No. 08-785
  • / Each
Description & Specifications
Specifications
Lid Yes
Format 24-well
Sterile No
Closure Yes
Pore Size 1μm
Surface Coating Fibrillar Collagen
For Use With (Application) Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption
No. of Wells 24
Culture Area 0.3cm2
Type HTS Caco 2 Assay System
Color Transparent
Includes 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid
Quantity 1/Cs.