Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
$938.55 - $4,182.31
Specifications
Pore Size 3μm
Type Membrane
For Use With (Application) Endothelial Cell Invasion
No. of Wells 24
View More Specs
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
08-774-378 Corning™
C354141
1/Pk. Pack for $938.55
08-774-379 Corning™
C354142
5/Pk. Pack of 5 Each for $4,182.31
Specifications
Description
  • Allows invasion of Human Microvascular Endothelial Cells (HMVEC-1) - a human microvascular endothelial cell line
  • Excludes invasion of 3T3 cells - a non-invasive fibroblast cell line
Description & Specifications
Specifications
3μm
Membrane
Endothelial Cell Invasion
24