Corning™ BioCoat™ 12mm #1 German Glass Coverslips, Round
Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ 12mm #1German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Manufacturer:  Corning™ C354089
 View more versions of this product
Catalog No. 08-774-387
Alert:
In October 2012, Corning acquired the majority of the Becton Dickinson Discovery Labware business. In 2015, the integration of the Biologics portion of the Discovery Labware brands (including Matrigel™, BioCoat™, FluoroBlok™ and PureCoat™) into their growing family of Corning Life Sciences brands will be complete. You will notice changes on their product and packaging artwork, but the products themselves and the catalog numbers will remain the same.
Specifications
Description
Coverslip, Slide; Corning; BioCoat; No. 1 German glass; Round; Collagen I; 22mm dia.; Bulk packed; 60/cs.
  • Optically clear, thin glass coverslips
  • Exhibits low background fluorescence—ideal for immunofluorescence microscopy
  • Constructed of No. 1 German glass; non-toxic to neurons
  • Each lot of cellware has been tested and found negative for bacteria and fungi
Fluorescence imaging of live cells, Confocal microscopy, High resolution and inverted microscopy, Phase contrast microscopy
Description & Specifications
Specifications
Slide Coverslip
No. 1 German Glass
22mm
Collagen I
0.472 in.
Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy