missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ BioCoat™ #1 German Glass Coverslips, Round

Coated with a variety of extracellular matrix substrates. Corning™ BioiCoat™ No. 1 German Glass Coverslips provide an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.

Supplier:  Corning™ 354088

 View more versions of this product

Catalog No. 08-774-386



Description

Description

Corning™ BioCoat™ German Glass coverslips with surface treatment (see individual Specification tables for coating). Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.

Shelf life: ≥ 3 months from date of shipment

Specifications

Specifications

Slide Coverslip
No. 1 German Glass
Fluorescence imaging of live cells, confocal microscopy, high resolution and inverted microscopy, phase contrast microscopy
60
Fibronectin
0.866 in.
22 mm
60
SDS
Documents

Documents

Promotions

Promotions