Please login to your online account to display your discounted pricing

Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant


Isolated from pituitary glands

Manufacturer: corning™  C356061

 View more versions of this product

Catalog No. CB-40060B

  • / Each
 

Description & Specifications

Specifications

Stability 3 Months
Lyophilized Yes
Cross Reactivity Human
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium
Source E. coli
Type Basic Fibroblast Growth Factor
Storage Requirements -20°C
Molecular Weight 17
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Purity >95% by SDS-PAGE
Quantity 10 x 10μg