Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant
Isolated from pituitary glands
Manufacturer:  Corning™ C356060
 View more versions of this product
Catalog No. CB-40060A
Description
Description
A broad range of highly purified natural and recombinant lymphokine and growth factor biochemicals designed specifically for use in basic cell biology research.
  • Products are tested for biological activity in appropriate assays, such as the ability of growth factors to stimulate proliferation or DNA synthesis
  • Lymphokines and growth factors are also sterile filtered generally using a 0.2μm membrane
  • Tested for the presence of bacteria, fungi and mycoplasma
Specification sheets include detailed information on quality control testing, product source, storage, stability and recommendations on proper experimental concentrations.
Order Info
  • FGF is supplied lyophilized, bFGF in 3.6mM Tris, PBS pH 7 buffer
  • bFGF (human) is lyophilized
Specifications
Specifications
Basic Fibroblast Growth Factor
Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Yes
>95% by SDS-PAGE
3 Months
E. coli
Human
Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium
17
5 x 10μg
-20°C
Documents