Please login to your online account to display your discounted pricing

Corning™ Basic Fibroblast Growth Factors (bFGF), Human Recombinant


Isolated from pituitary glands

Manufacturer: corning™  C354060

 View more versions of this product

Catalog No. CB-40060

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
For Use With (Application) Useful for the maintenance and differentiation of sympathetic and sensory neurons, as well as neuronal cells in-vitro
Stability 3 Months
Cross Reactivity Human
Lyophilized Yes
Purity >95% by SDS-PAGE
Molecular Weight 17
Formulation Lyophilized from phosphate buffered saline, carrier free, reconstitute in dH2O or serum-free medium
Source E. coli
Type Basic Fibroblast Growth Factor
Quantity 10μg