missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Bar Coded Storage Tubes: 1D/2D Bar Coded

For enhanced sample storage, identification and retrieval

Manufacturer:  Corning™ C8507

 View more versions of this product

Catalog No. 07-200-517



Description

Description

Feature a synchronized coding scheme that minimizes confusion and the risk of duplication, while enabling safe and secure sample identification.

  • Maximum Identification — Synchronized 2D and linear bar code
  • Maximum Information — 14 x 14 dot 2D bar code
  • Bar Code Stability — Laser-etched, not an attached label
  • Compound Compatibility — Polypropylene construction throughout creates inert vessel
  • Temp. tolerance: -180 to +110°C (screw cap); -80 to +110°C (TPE cap)
  • Length: 43.9mm
  • 96/rack

Specifications

Specifications

Thermoplastic Elastomer
960/Pk., 5Pk./Cs.
White
Polypropylene
43.9 mm
1D/2D Barcoded
Septum Cap
TPE Septum Cap
Autoclavable
96 1D/2D Bar Coded tubes, Enhanced sample storage, identification and retrieval
Mat
Non-sterile
SDS
Documents