Please login to your online account to display your discounted pricing

Corning™ 96-Well Special Optics Low Fluorescence Assay Plates


Ideal for spectroscopy, polarization, and imaging

Manufacturer: corning™  C3720

 View more versions of this product

Catalog No. 07200805

  • / Case of 25 Each
 

Description & Specifications

Specifications

For Use With (Application) Spectroscopy, polarization and imaging
Material Polystyrene
Quantity 5/Pk.
Sterile Yes
Type 96-Well Assay Microplate (Tissue culture treated)
Volume (Metric) Well 100 to 200μL
Volume (Metric) Working 75 to 200μL
Color Black
Applications Spectroscopy, polarization and imaging