Please login to your online account to display your discounted pricing

Corning™ 96-Well Special Optics Low Fluorescence Assay Plates

Ideal for spectroscopy, polarization, and imaging

$669.36 - $2,545.05

Specifications

For Use With (Application) Spectroscopy, polarization and imaging
Color Black
Material Polystyrene
Volume (Metric) Working 75 to 200μL
View More Specs
Products
Catalog Number Mfr. No. Quantity Sterile Type Price Quantity    

07200805

 
corning™
C3720
5/Pk. Yes 96-Well Assay Microplate (Tissue culture treated) Case of 25 Each for $669.36

07-200-729

 
corning™
C3614
25/Pk. Yes 96-Well Assay Microplate (Tissue culture treated) Case of 100 Each for $2,545.05

07-200-730

 
corning™
C3615
25/Pk. No 96-Well Assay Microplate (Untreated) Case of 100 Each for $2,528.56
Description & Specifications

Specifications

For Use With (Application) Spectroscopy, polarization and imaging
Color Black
Material Polystyrene
Volume (Metric) Working 75 to 200μL

  • Clear well bottoms are 60% thinner than conventional plates for direct viewing from either top or bottom
  • Black opaque walls emit minimal background for sharp, clear images of fluorescent cells
  • Available untreated or with tissue culture-treated surfaces

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.