missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 96-Well Nonbinding Surface (NBS™) Microplates

Treated with a Corning proprietary treatment technology to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Corning™ 96-Well Nonbinding Surface (NBS™) Microplates are ideal for reducing proteing and nucleic acid binding at low concentrations, and increasing assay signal to noise.

Manufacturer:  Corning™ C3600

 View more versions of this product

Catalog No. 07-200-731



Description

Description

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications

    Specifications

    Standard
    360 μL
    Flat
    White
    Plate readers, liquid handlers and robotic systems
    100/Cs.
    39°F to 99°F
    25/Pk.
    White
    96
    Without Lid
    75 to 200 μL
    Flat
    For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
    Polystyrene
    100/Cs.
    4° to 37°C
    Non-sterile
    Non-binding
    8 x 12
    SDS
    Documents