Corning™ 96-Well Half Area Flat Bottom Polystyrene NBS™ Microplates
Designed to reduce well-to-well crosstalk during luminescent assays
Manufacturer:  Corning™ C3642
 View more versions of this product
Catalog No. 07-201-201
Description
Description
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • Specifications
    Specifications
    Flat
    Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    100/Cs.
    25/Pk.
    Non-binding
    8 x 12
    25 to 125 μL
    Flat
    White
    Polystyrene
    100/Cs.
    Non-sterile
    96
    Without Lid
    Half-Area