Corning™ 96-Well Half Area Black Flat Bottom Polystyrene Microplates
Designed to reduce well-to-well crosstalk
$505.18 - $505.18
Specifications
For Use With (Application) Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
Material Polystyrene
Type 96-Well Assay Microplate
Volume (Metric) Well 190μL
Volume (Metric) Working 25 to 125μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Sterile Surface Coating Price Quantity  
Catalog Number Mfr. No. Color Sterile Surface Coating Price Quantity  
07-201-205 Corning™
C3993
Black No NBS Treated Case of 25 Each for $505.18
Specifications
Description
  • Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
  • Choice of NBS™ or TC-treated surface coating
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
96-Well Assay Microplate
25 to 125μL
Polystyrene
190μL
Flat (F)