missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 96-Well Clear-Bottom Half-Area Microplates

Designed with optically clear flat bottom that permits direct microscopic viewing. Corning™ 96-Well Clear Bottom Half Area Microplates provide an ideal solution for use in cell-based assays and microscopy applications allowing top or bottom reading capability.

$1,960.69 - $2,678.44

Specifications

Format 96-well half-area
Material Polystyrene
Volume (Metric) Well 190μL
For Use With (Equipment) Standard automated handling and detection systems
Volume (Metric) Working 25 to 125μL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Lid Sterile Surface Coating Price Quantity  
Catalog Number Mfr. No. Color Lid Sterile Surface Coating Price Quantity  
07-200-842 Corning™
C3883
White No Yes Non-Treated Case of 100 Each for $2,031.15
07-200-840 Corning™
C3880
Black No No Non-Treated Case of 100 Each for $1,960.69
07-200-843 Corning™
C3885
White Yes Yes Tissue Culture Case of 100 Each for $2,491.38
07-200-841 Corning™
C3882
Black Yes Yes Tissue Culture Case of 100 Each for $2,224.77
07-200-845 Corning™
C3884
White No No NBS™ Case of 100 Each for $2,678.44
07-200-844 Corning™
C3881
Black No No NBS™ Case of 100 Each for $2,556.90
Specifications

Description

 

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background flourescence and enabling readings down to 340nm
  • Polystyrene
  • Flat wells
  • Surface area per well: 0.16cm2
  • Alphanumeric grid
  • Opaque walls prevent well-to-well crosstalk
  • Compatible with standard automated handling and detection systems
  • Choice of three surfaces: untreated for homogenous assays, non-binding surface (NBS™) for the minimization of protein and nucleic acid binding, or tissue culture treated for cell growth

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

96-well half-area
190μL
25 to 125μL
Polystyrene
Standard automated handling and detection systems
Clear