missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
Color:White, Clear Bottom; Sterility:Sterile; Lid:With Lid
Supplier: Corning™ 3885
Description
Specifications
Half-Area | |
0.16 cm2 | |
White, Clear Bottom | |
Polystyrene | |
25 to 125 μL | |
Cell Culture | |
Homogenous and cell based assays | |
20/Pk. | |
Clear |
With Lid | |
8 x 12 | |
Standard automated handling and detection systems | |
96 | |
Sterile | |
Flat Bottom (F-well) | |
100/Cs. | |
100/Cs. |