Please login to your online account to display your discounted pricing

Corning™ 96-Well Clear-Bottom Half-Area Microplates


Designed with optically clear flat bottom that permits direct microscopic viewing. Corning™ 96-Well Clear Bottom Half Area Microplates provide an ideal solution for use in cell-based assays and microscopy applications allowing top or bottom reading capability.

Manufacturer: corning™  C3887

 View more versions of this product

Catalog No. 07-200-847

  • / Case of 100 Each
 

Description & Specifications

Specifications

Lid No
For Use With (Application) Homogenous and cell based assays
For Use With (Equipment) Standard automated handling and detection systems
Format 96-well half-area
Material Polystyrene
No. of Wells 96
Sterile Yes
Surface Area (Metric) 0.16cm2
Surface Coating Tissue Culture
Volume (Metric) Well 190μL
Volume (Metric) Working 25 to 125μL
Well Color Clear
Closure No
Color Black
Applications Homogenous and cell based assays
Well Color Clear
Wells 96

 

  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background flourescence and enabling readings down to 340nm
  • Polystyrene
  • Flat wells
  • Surface area per well: 0.16cm2
  • Alphanumeric grid
  • Opaque walls prevent well-to-well crosstalk
  • Compatible with standard automated handling and detection systems
  • Choice of three surfaces: untreated for homogenous assays, non-binding surface (NBS™) for the minimization of protein and nucleic acid binding, or tissue culture treated for cell growth