missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 50mL Easy ID Color Strip Polypropylene Centrifuge Tube

Easily identify samples contained in clear, color strip polypropylene tubes that eliminate sticker labeling

$481.13 - $663.33

Specifications

Material Polypropylene
Max. RCF 15,500xg
Length (Metric) 114.4mm
Shape Conical
Diameter (Metric) Outside 29.1mm
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Label Type Price Quantity  
Catalog Number Mfr. No. Label Type Price Quantity  
07-201-990 Corning™
C431525
Hazardous material band Case of 300 Each for $481.13
07-201-991 Corning™
C431526
Green band Case of 300 Each for $663.33
07-201-993 Corning™
C431528
Blue band Case of 300 Each for $663.33
07-201-992 Corning™
C431527
Orange band Case of 300 Each for $663.33
Specifications

Description

  • Conical bottom
  • Plug seal
  • Maximum RCF is 15,500xg
  • Sterile, nonpyrogenic, and certified RNase-/DNase-free
  • 95 k PA (14 psi) pressure tested
  • Recommended working temperature: 0-40°C
  • Heat distortion point: 121°C
Description & Specifications

Specifications

Polypropylene
114.4mm
29.1mm
15,500xg
Conical