Please login to your online account to display your discounted pricing

Corning™ 50mL Easy ID Color Strip Polypropylene Centrifuge Tube


Easily identify samples contained in clear, color strip polypropylene tubes that eliminate sticker labeling

Manufacturer: corning™  C431525

 View more versions of this product

Catalog No. 07-201-990

  • / Case of 300 Each
 

Description & Specifications

Specifications

Length (Metric) 114.4mm
Closure Type Plug seal cap
Max. RCF 15,500xg
Label Type Hazardous material band
For Use With (Application) Safe for air shipping
Capacity 50mL
Closure Color Orange
Disposable Yes
Packaging Rack packed
Purity Nonpyrogenic, and certified RNase-/DNase-free
Graduated Yes
Shape Conical
Diameter (Metric) Outside 29.1mm
Material Polypropylene
Graduations 5-50mL

  • Conical bottom
  • Plug seal
  • Maximum RCF is 15,500xg
  • Sterile, nonpyrogenic, and certified RNase-/DNase-free
  • 95 k PA (14 psi) pressure tested
  • Recommended working temperature: 0-40°C
  • Heat distortion point: 121°C