Please login to your online account to display your discounted pricing

Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates


Manufactured from medical grade polystyrene for strength. Corning™ 384-Well Clear Bottom Black or White Polystyrene Microplates permit top or bottom analysis using detection microplate readers.

Manufacturer: corning™  C3707

 View more versions of this product

Catalog No. 07-200-650

  • / Case of 100 Each
 

Description & Specifications

Specifications

Bottom Flat bottom
Volume (Metric) Well 112μL
Type 384-Well TC-treated Microplate
For Use With (Application) Attachment and growth of anchorage-dependent cells
Binding Property TC-treated
Lid Yes
Format 384-well
Material Polystyrene
Volume (Metric) 112μL
Closure Yes
Well Color Clear
Culture Area 0.06cm2
No. of Wells 384
Color White
Sterile Yes
Volume (Metric) Working 20 to 80μL

 

  • Molded polystyrene with opaque walls and optically clear bottoms
  • Readable from top or bottom for direct microscope viewing and CCD camera-based detectors
  • Opaque black or white walls prevent well to well crosstalk
  • Recommended working volumes of 20 to 80μL; total well volume of 112μL
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Suited for flourescent and luminescent assays
  • Plate dimensions meet proposed industry standard