missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ 384-Well Optical Imaging Microplates

Ideal for whole cell or subcellular high content screening

$1,681.59 - $4,780.41

Specifications

Culture Area 0.05cm2
Color Black
Sterile Yes
Format 384-well
Material Polystyrene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Volume (Metric) Working Lid Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Includes Volume (Metric) Working Lid Volume (Metric) Price Quantity  
07-201-01 Corning™
C3985BC
With generic barcode 20 to 80μL Yes 112μL Case of 100 Each for $4,780.41
07-201-11 Corning™
C3821BC
With generic barcode 5 to 40μL No 50μL Case of 50 Each for $1,681.59
07-200-850 Corning™
C3985
Without barcode 20 to 80μL Yes 112μL Case of 100 Each for $4,541.82
Specifications

Description

  • Compatible with high throughput confocal imagers
  • Better cell growth and attachment than glass bottom plates
  • TC-treated for optimal cell attachment

Generic Barcode

  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all 4 sides of the plate
  • No sequence number duplicates

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

0.05cm2
Yes
Polystyrene
Clear
Black
384-well
384-Well Optical Imaging Microplate
384