Please login to your online account to display your discounted pricing

Corning™ 384-Well Nonbinding Surface (NBS™) Microplates


Enhanced signal-to-noise ratio

Manufacturer: corning™  C3653

 View more versions of this product

Catalog No. 07-201-105

  • / Case of 100 Each
 

Description & Specifications

Specifications

Color White
For Use With (Application) For homogenous assays such as scintillation proximity assays (SPA) and fluorescent assays
For Use With (Equipment) Plate readers, liquid handlers and robotic systems
Format 384-well
Sterile No
Surface Coating NBS™
Temperature Range (English) 39° to 99°F
Temperature Range (Metric) 4° to 37°C
Volume (Metric) Well 112μL
Volume (Metric) Working 20 to 80μL
Well Color Clear