missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ Untreated 245mm Square BioAssay Dishes

Designed with a generous growth area of 500cm2 to allow for larger assays and better overall use of space in incubators. Corning™ Untreated 245mm Square Low Profile BioAssay Dishes provide a superior option for applications where cell attachment is not desired.

Manufacturer:  Corning™ C431272

 View more versions of this product

Catalog No. 06-443-22



Description

Description

Improve the efficiency of microbial assays including library replication, colony picking, and plaque lift studies.

 

  • Polystyrene dishes feature stacking beads for stable stacking
  • Compatible with various automated colony picking and gridding robots
  • Supplied with vented lid to provide consistent gas exchange
  • Sterile, certified nonpyrogenic
  • Patented (U.S. Patent No.. 5,856,176) design allows for easier handling

 

Specifications

Specifications

Clear
25 mm
Clear
Closure Included
With handles
Sterile
Cell Culture Dish
150 mL (Well)
245 mm
Square
Square
Polystyrene
500 sq. cm
245 x 245 mm
Polystyrene
Disposable
Lid
245 mm
SDS
Documents
Product Certifications