Corning™ 1536-Well Tissue Culture-Treated Polystyrene Plates
Features excellent signal dynamic range due to low background fluorescence and enhanced signal intensity. Corning™1536-Well Tissue Culture-Treated Polystyrene Plates provide superior flatness which minimizes variability within a plate and between plates.
$3,250.97 - $7,217.56
Specifications
Culture Area 0.03cm2
Sterile Yes
Material Polystyrene
Volume (Metric) 12.5μL
Volume (Metric) Working 8μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Color Lid Well Color Price Quantity  
Catalog Number Mfr. No. Color Lid Well Color Price Quantity  
07-201-08 Corning™
C3727BC
White Yes White Case of 50 Each for $3,250.97
07-202-513 Corning™
C3727
White Yes White Case of 50 Each for $3,334.41
12-456-701 Corning™
C4561
Black No Clear Case of 100 Each for $7,217.56
12-456-705 Corning™
C4566
Black No Black Case of 50 Each for $4,314.62
12-456-709 Corning™
C4571
White No White Case of 50 Each for $4,314.62
07-202-512 Corning Incorporated
C3726
Black Yes Black Case of 50 Each for $3,379.90
Specifications
Description
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™
  • Working volume of up to 10μL
  • Available untreated, with nonbinding surface (NBS™), or tissue culture treated
  • NBS microplates are ideal for homogeneous assays with “sticky” enzymes or for reducing nonspecific binding to microplate wells
Generic Barcode
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate
  • No sequence number duplicates
Order Info
Packaged in bags of ten
Description & Specifications
Specifications
0.03cm2
Polystyrene
8μL
Yes
12.5μL
1536