Please login to your online account to display your discounted pricing

1536-Well Polystyrene NBS™-Treated Microplates


Proprietary nonbinding surface is ideal for homogeneous assays with sticky enzymes or for reducing nonspecific binding to microplate wells

Manufacturer: corning incorporated  7429

 View more versions of this product

Catalog No. MRK7429

  • / Case of 50 Each
 

Description & Specifications

Specifications

Closure No
Color Not Applicable
Format 1536-well
Includes Without barcode
Material Cyclic Olefin copolymer
No. of Wells 1536
Quantity 50/Cs.
Sterile No
Type 1536-Well Microplate
Volume (Metric) N/A
Volume (Metric) Working N/A
Well Color N/A
Lid No

  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Superior flatness minimizes variability within a plate and between plates
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer FlexDrop™

Generic Barcode

  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate
  • No sequence number duplicates