Please login to your online account to display your discounted pricing
Corning™ 1536-Well CellBIND™ Surface-Treated Polystyrene Microplates
$4,885.77 - $5,202.56
Specifications
Lid No
Coding Barcode
Material Cyclic Olefin Copolymer
Sterile Yes
Surface Coating Corning™ Cellbind™
View More Specs
Products
Catalog Number Mfr. No. Quantity Color Price Quantity    
12-456-707
corning™
C4568
10/Pk. Black Case of 50 Each for $4,885.77
12-456-711
corning™
C4573
10/Pk. White Case of 50 Each for $5,202.56
Description & Specifications
Specifications
Lid No
Coding Barcode
Material Cyclic Olefin Copolymer
Sterile Yes
Surface Coating Corning™ Cellbind™
Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers
Volume (Metric) Well 5 to 8μL
Volume (Metric) Working 8μL