Please login to your online account to display your discounted pricing

Corning™ 1536-Well CellBIND™ Surface-Treated Polystyrene Microplates


Manufacturer: corning™  C4573

 View more versions of this product

Catalog No. 12-456-711

  • / Case of 50 Each
 

Description & Specifications

Specifications

Lid No
Sterile Yes
Closure No
Material Cyclic Olefin Copolymer
Volume (Metric) Working 8μL
Type 1536-Well Assay Microplate
Quantity 10/Pk.
For Use With (Application) For higher sensitivity
For Use With (Equipment) Automated dispensers
Volume (Metric) Well 5 to 8μL
Color White
Surface Coating Corning™ Cellbind™
Coding Barcode

  • Smooth top; Clear flat bottom; Low base
  • No logo or lettering
  • With barcode
  • Without lid