Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates
Highest density microplates available for high throughput screening
$5,086.56 - $7,506.64
Specifications
Lid No
Volume (Metric) Working 8μL
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Color Surface Coating Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Color Surface Coating Type Price Quantity  
05-539-30 Corning™
C3891
For fluorescent homogeneous assays, high throughput screening Black, Clear Non-Treated 1536-Well Assay Microplate Case of 50 Each for $5,086.56
07-200-872 Corning™
C3895BC
For fluorescent homogeneous assay Black, Clear NBS™ 1536-Well Assay Microplate Case of 50 Each for $7,506.64
13-701-101 Corning™
C3839BC
For fluorescent and luminescent assays Black Poly-D-Lysine 1536-Well Multiwell Plate N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Description
Conform to standard microplate footprint and dimensions. 2μLwell microplate is a thinner plate with the length and width dimensions and microplate footprint meeting industry standards.
  • For fluorescent and luminescent assays
  • Total well volume of 10μL for round well plates and 12.8μL for flat bottom plates
  • Recommended working volume of up to 8μL
  • Round well bottoms for reduced air entrapment and improved CVs and Z factor
  • Raised well bottoms for higher sensitivity
  • Flood reservoir on four sides to reduce instrument contamination
  • Flat well bottoms
Order Info
Packaged in bags of 10
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Description & Specifications
Specifications
No
8μL