Please login to your online account to display your discounted pricing

Corning™ 1536-Well Black Clear-Bottom Polystyrene Plates


Highest density microplates available for high throughput screening

Manufacturer: corning™  C3895BC

 View more versions of this product

Catalog No. 07-200-872

  • / Case of 50 Each
 

Description & Specifications

Specifications

Lid No
For Use With (Application) For fluorescent homogeneous assay
Material Polystyrene
Bottom Flat (F)
Sterile No
Surface Coating NBS™
Type 1536-Well Assay Microplate
Volume (Metric) Well 12.5μL
Volume (Metric) Working 8μL
Closure No
Color Black, Clear
Closure No
Volume (Metric) Well 12.5μL
Applications For fluorescent homogeneous assay
Well Bottom Flat (F)