Please login to your online account to display your discounted pricing

Corning™ 0.05% Trypsin/0.53mM EDTA in HBSS w/o Calcium, Magenesium or Sodium Bicarbonate


Used to enzymatically release adherent cells from tissue culture plates for passaging.

Manufacturer: corning™  25052CI

 View more versions of this product

Catalog No. MT25052CI

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Sterile Yes
Without Additives Calcium, Magnesium, Sodium Bicarbonate
Solution Type 0.05% Trypsin/0.53mM EDTA in HBSS
Storage Requirements -20°C
Quantity 6 × 100mL
Format Liquid
pH 7 to 7.6

    • EDTA enhances the efficacy of trypsin
    • Manufactured under industry-leading standards for low endotoxin content and superior results
    • Certificate of Analysis available
    • Enzymatic dissociation reagents tested for porcine parvovirus