Please login to your online account to display your discounted pricing

Cornerstones of Chemistry: The Mole


Poster offers graphic depiction of the mole and its importance

Manufacturer: fisher science education  AZ-35-1160

 View more versions of this product

Catalog No. S90659A

  • / Each
 

Description & Specifications

Specifications

Product Type Cornerstones of Chemistry - the Mole Poster
Classification Science
For Use With (Application) Provides a clear and interesting presentation of Avogadro's law, the uniqueness of the mole, molarity, molality, molecular formulas
Grade 7 to 12
Height (English) 35 in.
Height (Metric) 88.9cm
Includes Illustrated teacher's guide
Material High-Gloss Paper
Width (English) 23 in.
Width (Metric) 58.42cm