Please login to your online account to display your discounted pricing

Cornerstones of Chemistry: The Mole

Poster offers graphic depiction of the mole and its importance

$16.20 - $16.20

Specifications

Classification Science
For Use With (Application) Provides a clear and interesting presentation of Avogadro's law, the uniqueness of the mole, molarity, molality, molecular formulas
Grade 7 to 12
Height (English) 35 in.
Height (Metric) 88.9cm
View More Specs
Includes:  Illustrated teacher's guide
Products
Catalog Number Mfr. No. Material Price Quantity    

S90659A

 
fisher science education
AZ-35-1160
High-Gloss Paper Each for $16.20
Description & Specifications

Specifications

Classification Science
For Use With (Application) Provides a clear and interesting presentation of Avogadro's law, the uniqueness of the mole, molarity, molality, molecular formulas
Grade 7 to 12
Height (English) 35 in.
Height (Metric) 88.9cm
Includes Illustrated teacher's guide
Width (English) 23 in.
Width (Metric) 58.42cm

Provides a clear and interesting presentation of:

  • Avogadro's law
  • Uniqueness of the mole
  • Molarity
  • Molality
  • Molecular formulas
  • Everyday uses of the mole in science and industry

Grades 6-12. Dimensions (L x W): 35 x 25 in. (89 x 63.5cm).