Please login to your online account to display your discounted pricing

Corks of Assorted Sizes


Manufacturer: fisher science education  12-26ASST

 View more versions of this product

Catalog No. S50028

  • / Pack of 100 Each
 

Description & Specifications

Specifications

Color Natural
Size 12 to 16
Stopper Number Assorted
Material Cork
For Use With (Application) For sealing bottles, especially for wine bottles
Product Type Stopper