Cork Borer Sets with Handles
Bore holes in corks and rubber stoppers.
Manufacturer: Fisher Science Education H9665
Description
- Consist of plated tubing with precision-ground cutting edges
- Smooth plastic handles
- Handles are stamped with a number designation
- Punch included with each set
Specifications
Cork Borer | |
4.8 to 23.8mm | |
Plated tubing, smooth Plastic handles |
0.18 to 0.93 in. | |
To bore holes in corks and Rubber stoppers | |
15 Pack |