Please login to your online account to display your discounted pricing

(-)-Corey lactone 4-phenylbenzoate alcohol, 99%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  224502500

 View more versions of this product

Catalog No. AC224502500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Assay ≥98.5%
Formula Weight 352.38
Quantity 250mg
Color White to off-white
TSCA TSCA
Melting Point 128° to 134°C
CAS Max % 100.0
Name Note 99%
Synonym (3aR,4S,5R,6aS)-(-)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1'-biphenyl-4-carboxylate
Specific Rotation -84° to -89° (20°C, 589nm) (c=1, CHCl3)
Chemical Name or Material (-)-Corey lactone 4-phenylbenzoate alcohol
Assay Percent Range 99%
Molecular Formula C21H20O5
MDL Number MFCD00078077
CAS 31752-99-5
CAS Min % 98.5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material (-)-Corey lactone 4-phenylbenzoate alcohol
Name Note 99%
CAS 31752-99-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C21H20O5
Formula Weight 352.38
MDL Number MFCD00078077
Synonym (3aR,4S,5R,6aS)-(-)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1'-biphenyl-4-carboxylate