missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(-)-Corey lactone 4-phenylbenzoate alcohol, 99%, ACROS Organics™

Chemical Identifiers

CAS 31752-99-5
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C21H20O5
Formula Weight 352.38
MDL Number MFCD00078077
Synonym (3aR,4S,5R,6aS)-(-)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1'-biphenyl-4-carboxylate
Chemical Name or Material (-)-Corey lactone 4-phenylbenzoate alcohol, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC224502500 SDS Thermo Fisher Scientific
224502500
Glass bottle 250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

31752-99-5
100.0
C21H20O5
MFCD00078077
(-)-Corey lactone 4-phenylbenzoate alcohol, 99%
98.5
99%
352.38
(3aR,4S,5R,6aS)-(-)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1'-biphenyl-4-carboxylate
≥98.5%

Specifications

352.38
250mg
128° to 134°C
(3aR,4S,5R,6aS)-(-)-Hexahydro-4-(hydroxymethyl)-2-oxo-2H-cyclopenta[b]furan-5-yl 1,1'-biphenyl-4-carboxylate
31752-99-5
MFCD00078077
99%
Authentic
≥98.5%
Fine Crystalline Powder
White to off-white
100.0
-84° to -89° (20°C, 589nm) (c=1, CHCl3)
C21H20O5
98.5
Glass bottle
(-)-Corey lactone 4-phenylbenzoate alcohol, 99%

Safety and Handling

TSCA : TSCA