Please login to your online account to display your discounted pricing

Alfa Aesar™ Corazonin


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66997LB0

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66997LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 5mg
Format Solid
Sequence Glp-Thr-Phe-Gln-Tyr-Ser-Arg-Gly-Trp-Thr-Asn-NH2
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type Corazonin
Molecular Weight 1369.46
Purification Method HPLC