Please login to your online account to display your discounted pricing

COR 170, Tocris™

Selective CB2 receptor inverse agonist

$246.52 - $1,023.37

Chemical Identifiers

Chemical Name or Material COR 170
Assay Percent Range >99%
Molecular Formula C31H36N2O2
Formula Weight 468.63
Synonym N-(Adamant-1-yl)-4-oxo-1-pentyl-6-phenyl-1, 4-dihydroquinoline-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

475650

 
r & d systems
RDS475650
50mg Each for $1,023.37

475610

 
r & d systems
RDS475610
10mg Each for $246.52
Description & Specifications

Specifications

Formula Weight 468.63
Quantity 50mg
Chemical Name or Material COR 170
CAS 1048039-15-1
Synonym N-(Adamant-1-yl)-4-oxo-1-pentyl-6-phenyl-1, 4-dihydroquinoline-3-carboxamide
Molecular Formula C31H36N2O2
Assay Percent Range >99%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature