missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper wire, 99.9985% (metals basis), Alfa Aesar™ Puratronic™

Form: approx. 0.44g/m

Manufacturer:  Thermo Fisher Scientific 10972G6

 View more versions of this product

Catalog No. AA10972G6



Specifications

Specifications

Copper
7440-50-8
8.94g/cm3
Cu
anode
[Cu]
Odorless
23978
Wire, 0.25mm (0.01 in.) dia.
Undesignated
10m
Copper colored wire
99.9985% (Metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
0 hPa

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

Documents