missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Copper Wire, 2.0mm (0.08 in.) dia, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Form: Approx. 28.1g/m

Manufacturer:  Alfa Aesar 10975BW

 View more versions of this product

Catalog No. AA10975BW



Specifications

Specifications

Copper wire
2.0mm (0.08 in.) dia, Puratronic™
≈28.1g/m
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
99.999%
50cm
7440-50-8
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents