Please login to your online account to display your discounted pricing

Copper wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Copper wire
CAS 7440-50-8
Name Note 2.0mm (0.08 in.) dia, Puratronic™
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA10975BW

  SDS
alfa aesar
10975BW
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50cm
Chemical Name or Material Copper wire
TSCA TSCA
Density ≈28.1g/m
Name Note 2.0mm (0.08 in.) dia, Puratronic™
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
Odor Odorless