missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper wire, 1.5mm (0.06 in.) dia., Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 44551G5

 View more versions of this product

Catalog No. AA44551G5



Specifications

Specifications

Copper wire
5m
7440-50-8
Cu
anode
[Cu]
Odorless
23978
Wire, ≈15.8g/m
Puratronic™
≈15.8g/m
1.5mm (0.06 in.) dia.
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.9999%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

EINECSNumber : 231-159-6

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents