missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper wire, 1.0mm (0.04 in.) dia., Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 10974G1

 View more versions of this product

Catalog No. AA10974G1



Specifications

Specifications

Copper wire
1m
7440-50-8
Cu
anode
[Cu]
Odorless
23978
Wire
Puratronic™
≈7.02g/m
1.0mm (0.04 in.) dia.
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.999%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
anode
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents