Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper wire, 0.5mm (0.02 in.) dia., Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Copper wire
CAS 7440-50-8
Name Note 0.5mm (0.02 in.) dia.
Purity Grade Puratronic™
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA10973G5

 
alfa aesar™
10973G5
5m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10973H4

 
alfa aesar™
10973H4
100m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10973G9

 
alfa aesar™
10973G9
25m CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper wire
TSCA TSCA
Name Note 0.5mm (0.02 in.) dia.
Purity Grade Puratronic™
Physical Form Wire
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
Odor Odorless
Weight ≈1.75g/m
Quantity 5m