missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper wire, 0.5mm (0.02 in.) dia., Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 10973G9

 View more versions of this product

Catalog No. AA10973G9



Specifications

Specifications

Copper wire
25m
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
Wire
Puratronic™
≈1.75g/m
0.5mm (0.02 in.) dia.
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
99.999%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents