missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper tubing, 3.175mm (0.125in) OD, 1.5mm (0.059in) ID, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 13471RM

 View more versions of this product

Catalog No. AA13471RM



Specifications

Specifications

Copper tubing
OD: 3.175mm (0.125 in.) ID: 1.5mm (0.059 in.)
(metals basis)
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
copper
63.546
CHEBI:30052
90cm
7440-50-8
Cu
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
[Cu]
Odorless
23978
99.9%

Chemical Identifiers

7440-50-8
63.546
RYGMFSIKBFXOCR-UHFFFAOYSA-N
23978
copper
Cu
MFCD00010965
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
CHEBI:30052
[Cu]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents