Please login to your online account to display your discounted pricing

Alfa Aesar™ Copper tubing, 1.59mm (0.0626in) OD, 0.876mm (0.0345in) ID, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Copper tubing
CAS 7440-50-8
Name Note OD: 1.59mm (0.0626 in.) ID: 0.876mm (0.0345 in.)
MDL Number MFCD00010965
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13474RM

 
alfa aesar™
13474RM
90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13474RN

 
alfa aesar™
13474RN
5 x 90cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Copper tubing
TSCA TSCA
Name Note OD: 1.59mm (0.0626 in.) ID: 0.876mm (0.0345 in.)
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD00010965
CAS 7440-50-8
Odor Odorless
Quantity 90cm